Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITIH4
Entrez Gene ID
3700
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Gene Aliases
GP120, H4P, IHRP, ITI-HC4, ITIHL1, PK-120, PK120
Chromosome Location
Chr.3: 52812990 - 52830701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166449.1 NP_001159921.1 3 - 4 411 115
NM_002218.4 NP_002209.2 3 - 4 411 115
GenBank mRNA AB209388.1 - 2 - 3 1543 115
AK298407.1 - - 382 115
AK298412.1 - 3 - 4 382 115
AK298429.1 - 3 - 4 382 115
AK303769.1 - 2 - 3 237 115
BC136392.1 - 3 - 4 383 115
BC136393.1 - 3 - 4 383 115
BX477542.1 - 3 - 4 389 115
D38595.1 - 3 - 4 115
DC412911.1 - 3 - 4 413 115

View Full Product Details