The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204760.1 | NP_001191689.1 | 5 - 6 | 1279 | 110 |
NM_012464.4 | NP_036596.3 | 5 - 6 | 1279 | 110 | |
XM_011532214.1 | XP_011530516.1 | 5 - 6 | 1337 | 110 | |
XM_017008570.1 | XP_016864059.1 | 6 - 7 | 1405 | 110 | |
GenBank mRNA | AF282732.1 | - | 5 - 6 | 883 | 110 |
BC016922.2 | - | 5 - 6 | 797 | 110 | |
BC136429.1 | - | 5 - 6 | 1189 | 110 | |
BC136430.1 | - | 5 - 6 | 1189 | 110 | |
BC144085.1 | - | 5 - 6 | 1189 | 110 | |
U91963.1 | - | 5 - 6 | 1279 | 110 |