The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204760.1 | NP_001191689.1 | 8 - 9 | 1690 | 69 |
NM_012464.4 | NP_036596.3 | 8 - 9 | 1690 | 69 | |
XM_011532214.1 | XP_011530516.1 | 8 - 9 | 1748 | 69 | |
XM_017008570.1 | XP_016864059.1 | 9 - 10 | 1816 | 69 | |
GenBank mRNA | AF282732.1 | - | 8 - 9 | 1294 | 69 |
BC016922.2 | - | 8 - 9 | 1208 | 69 | |
BC136429.1 | - | 8 - 9 | 1600 | 69 | |
BC136430.1 | - | 8 - 9 | 1600 | 69 | |
BC144085.1 | - | 8 - 9 | 1600 | 69 | |
U91963.1 | - | 8 - 9 | 1690 | 69 |