The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000790.3 | NP_000781.1 | 14 - 15 | 1548 | 111 |
NM_001082971.1 | NP_001076440.1 | 14 - 15 | 1663 | 111 | |
NM_001242886.1 | NP_001229815.1 | 13 - 14 | 1434 | 111 | |
NM_001242887.1 | NP_001229816.1 | 13 - 14 | 1404 | 111 | |
NM_001242888.1 | NP_001229817.1 | 12 - 13 | 1314 | 111 | |
NM_001242889.1 | NP_001229818.1 | 12 - 13 | 1269 | 111 | |
XM_005271745.4 | XP_005271802.1 | 13 - 14 | 1552 | 111 | |
XM_011515161.2 | XP_011513463.2 | 12 - 13 | 1420 | 111 | |
GenBank mRNA | AK298321.1 | - | 13 - 14 | 1379 | 111 |
AK298392.1 | - | 13 - 14 | 1424 | 111 | |
BC000485.1 | - | 14 - 15 | 1532 | 111 | |
BC008366.1 | - | 14 - 15 | 1537 | 111 | |
BM713211.1 | - | 1 - 2 | 51 | 111 | |
CD013934.1 | - | 12 - 13 | 1297 | 111 | |
CD013935.1 | - | 12 - 13 | 1252 | 111 | |
CD013936.1 | - | 13 - 14 | 1417 | 111 | |
CD013937.1 | - | 13 - 14 | 1387 | 111 | |
CD013938.1 | - | 12 - 13 | 1322 | 111 | |
M76180.1 | - | 14 - 15 | 1531 | 111 | |
M88700.1 | - | 14 - 15 | 1551 | 111 |