Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
Chromosome Location
Chr.9: 33252471 - 33264761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KU177933.1 - 3 - 4 510 118

View Full Product Details