The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021198.2 | NP_067021.1 | 2 - 3 | 555 | 65 |
XM_011511569.2 | XP_011509871.1 | 2 - 3 | 702 | 65 | |
XM_017004615.1 | XP_016860104.1 | 2 - 3 | 713 | 65 | |
XM_017004617.1 | XP_016860106.1 | 2 - 3 | 1137 | 65 | |
GenBank mRNA | AF229162.1 | - | 2 - 3 | 555 | 65 |
BC012977.2 | - | 2 - 3 | 264 | 65 |