The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206878.1 | NP_001193807.1 | 3 - 4 | 395 | 95 |
NM_021198.2 | NP_067021.1 | 3 - 4 | 659 | 95 | |
NM_182642.2 | NP_872580.1 | 3 - 4 | 656 | 95 | |
XM_011511569.2 | XP_011509871.1 | 3 - 4 | 806 | 95 | |
XM_011511570.2 | XP_011509872.1 | 3 - 4 | 802 | 95 | |
XM_017004615.1 | XP_016860104.1 | 3 - 4 | 817 | 95 | |
XM_017004616.1 | XP_016860105.1 | 3 - 4 | 812 | 95 | |
XM_017004617.1 | XP_016860106.1 | 3 - 4 | 1241 | 95 | |
XM_017004618.1 | XP_016860107.1 | 3 - 4 | 423 | 95 | |
GenBank mRNA | AF217969.1 | - | 1 - 2 | 63 | 95 |
AF229162.1 | - | 3 - 4 | 659 | 95 | |
AF258582.1 | - | 1 - 2 | 119 | 95 | |
AY279529.1 | - | 3 - 4 | 320 | 95 | |
AY279530.1 | - | 2 - 3 | 179 | 95 | |
BC012977.2 | - | 3 - 4 | 368 | 95 | |
BX424971.2 | - | 3 - 4 | 283 | 95 |