The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040427.1 | NP_001035517.1 | 3 - 4 | 747 | 71 |
NM_001286249.1 | NP_001273178.1 | 2 - 3 | 651 | 71 | |
NM_005326.4 | NP_005317.2 | 2 - 3 | 651 | 71 | |
XM_011522469.2 | XP_011520771.1 | 2 - 3 | 291 | 71 | |
XM_011522470.2 | XP_011520772.1 | 2 - 3 | 289 | 71 | |
XM_017023195.1 | XP_016878684.1 | 2 - 3 | 216 | 71 | |
GenBank mRNA | AK290155.1 | - | 2 - 3 | 216 | 71 |
AK298174.1 | - | 2 - 3 | 291 | 71 | |
AK299173.1 | - | 2 - 3 | 282 | 71 | |
AK299993.1 | - | 2 - 3 | 291 | 71 | |
AK308825.1 | - | 2 - 3 | 259 | 71 | |
BC000840.1 | - | 2 - 3 | 271 | 71 | |
BC002627.2 | - | 2 - 3 | 600 | 71 | |
BU176541.1 | - | 3 - 4 | 352 | 71 | |
DB502986.1 | - | 3 - 4 | 376 | 71 | |
X90999.1 | - | 1 - 2 | 136 | 71 |