The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040427.1 | NP_001035517.1 | 7 - 8 | 1145 | 70 |
NM_001286249.1 | NP_001273178.1 | 5 - 6 | 940 | 70 | |
NM_005326.4 | NP_005317.2 | 6 - 7 | 1049 | 70 | |
XM_011522469.2 | XP_011520771.1 | 6 - 7 | 689 | 70 | |
XM_011522470.2 | XP_011520772.1 | 5 - 6 | 578 | 70 | |
XM_017023195.1 | XP_016878684.1 | 6 - 7 | 614 | 70 | |
GenBank mRNA | AK298174.1 | - | 5 - 6 | 580 | 70 |
AK299173.1 | - | 6 - 7 | 680 | 70 | |
AK299993.1 | - | 6 - 7 | 680 | 70 | |
BC000840.1 | - | 6 - 7 | 669 | 70 | |
BC002627.2 | - | 6 - 7 | 998 | 70 | |
X90999.1 | - | 5 - 6 | 534 | 70 |