The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199125.1 | NP_001186054.1 | 11 - 12 | 1294 | 78 |
NM_001316354.1 | NP_001303283.1 | 11 - 12 | 1428 | 78 | |
NM_001316355.1 | NP_001303284.1 | 10 - 11 | 1167 | 78 | |
NM_001316356.1 | NP_001303285.1 | 10 - 11 | 1211 | 78 | |
NM_001316357.1 | NP_001303286.1 | 10 - 11 | 1291 | 78 | |
NM_001316358.1 | NP_001303287.1 | 9 - 10 | 1161 | 78 | |
NM_003653.3 | NP_003644.2 | 11 - 12 | 1341 | 78 | |
XM_005256837.4 | XP_005256894.1 | 11 - 12 | 1360 | 78 | |
XM_005256840.3 | XP_005256897.1 | 11 - 12 | 1405 | 78 | |
XM_005256842.4 | XP_005256899.1 | 10 - 11 | 1273 | 78 | |
XM_017025246.1 | XP_016880735.1 | 10 - 11 | 1199 | 78 | |
GenBank mRNA | AF031647.1 | - | 11 - 12 | 1246 | 78 |
AF098109.1 | - | 11 - 12 | 1266 | 78 | |
AK297706.1 | - | 11 - 12 | 1316 | 78 | |
AK302304.1 | - | 11 - 12 | 1294 | 78 | |
AK312476.1 | - | 11 - 12 | 1282 | 78 | |
AK316400.1 | - | 11 - 12 | 1373 | 78 | |
BC001891.1 | - | 11 - 12 | 1315 | 78 | |
BQ018146.1 | - | 1 - 2 | 78 | ||
BU952436.1 | - | 1 - 2 | 78 |