Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS3
Entrez Gene ID
8533
Gene Name
COP9 signalosome subunit 3
Gene Aliases
CSN3, SGN3
Chromosome Location
Chr.17: 17246624 - 17281303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316354.1 NP_001303283.1 3 - 4 439 134
XM_005256840.3 XP_005256897.1 3 - 4 416 134
XM_005256842.4 XP_005256899.1 2 - 3 284 134
GenBank mRNA AK297706.1 - 3 - 4 327 134

View Full Product Details