The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024701.3 | NP_078977.2 | 4 - 5 | 566 | 69 |
NR_024581.1 | 3 - 4 | 415 | 69 | ||
GenBank mRNA | AF403031.1 | - | 4 - 5 | 548 | 69 |
AK023196.1 | - | 4 - 5 | 549 | 69 | |
AK292071.1 | - | 4 - 5 | 552 | 69 | |
BG436652.1 | - | 3 - 4 | 392 | 69 | |
CR457302.1 | - | 4 - 5 | 522 | 69 |