Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP25
Entrez Gene ID
29761
Gene Name
ubiquitin specific peptidase 25
Gene Aliases
USP21
Chromosome Location
Chr.21: 15729947 - 15880072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283041.1 NP_001269970.1 13 - 14 1829 78
NM_001283042.1 NP_001269971.1 13 - 14 1829 78
NM_013396.4 NP_037528.3 13 - 14 1829 78
XM_005260949.4 XP_005261006.1 3 - 4 378 78
XM_006723992.3 XP_006724055.1 6 - 7 681 78
XM_011529529.1 XP_011527831.1 13 - 14 1907 78
XM_011529531.2 XP_011527833.1 12 - 13 1604 78
XM_011529533.2 XP_011527835.1 6 - 7 666 78
XM_011529535.2 XP_011527837.1 13 - 14 1907 78
XM_017028321.1 XP_016883810.1 13 - 14 1510 78
XM_017028322.1 XP_016883811.1 12 - 13 1783 78
XM_017028323.1 XP_016883812.1 11 - 12 1228 78
XM_017028324.1 XP_016883813.1 13 - 14 1681 78
XM_017028325.1 XP_016883814.1 13 - 14 1907 78
XM_017028327.1 XP_016883816.1 8 - 9 841 78
XM_017028328.1 XP_016883817.1 13 - 14 1907 78
XM_017028329.1 XP_016883818.1 8 - 9 841 78
GenBank mRNA AF134213.1 - 13 - 14 1659 78
AF170562.1 - 13 - 14 1829 78
AK022447.1 - 1 - 2 131 78
AK022574.1 - 4 - 5 512 78
BC075792.1 - 13 - 14 1830 78
FJ763652.1 - 13 - 14 1460 78
HG765100.1 - 3 - 4 362 78

View Full Product Details