The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006264.2 | NP_006255.1 | 13 - 14 | 2493 | 152 |
NM_080683.2 | NP_542414.1 | 13 - 14 | 2493 | 152 | |
NM_080684.2 | NP_542415.1 | 13 - 14 | 2493 | 152 | |
NM_080685.2 | NP_542416.1 | 13 - 14 | 2493 | 152 | |
XM_011532165.1 | XP_011530467.1 | 13 - 14 | 2501 | 152 | |
XM_017008511.1 | XP_016864000.1 | 13 - 14 | 2501 | 152 | |
XM_017008512.1 | XP_016864001.1 | 13 - 14 | 2501 | 152 | |
XM_017008513.1 | XP_016864002.1 | 13 - 14 | 2501 | 152 | |
XM_017008514.1 | XP_016864003.1 | 13 - 14 | 2501 | 152 | |
XM_017008515.1 | XP_016864004.1 | 12 - 13 | 1940 | 152 | |
XM_017008516.1 | XP_016864005.1 | 13 - 14 | 2501 | 152 | |
GenBank mRNA | AB208904.1 | - | 12 - 13 | 1917 | 152 |
BC140777.1 | - | 12 - 13 | 2032 | 152 | |
D21209.1 | - | 13 - 14 | 2076 | 152 | |
D21210.1 | - | 13 - 14 | 2076 | 152 | |
D21211.1 | - | 13 - 14 | 2076 | 152 | |
U12128.1 | - | 12 - 13 | 2230 | 152 | |
X80289.1 | - | 13 - 14 | 2090 | 152 |