Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL5
Entrez Gene ID
8869
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
SATI, SIAT9, SIATGM3S, ST3GalV
Chromosome Location
Chr.2: 85839148 - 85889034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042437.1 NP_001035902.1 2 - 3 197 135
NM_003896.3 NP_003887.3 2 - 3 342 135
XM_005264630.4 XP_005264687.1 2 - 3 298 135
XM_011533143.2 XP_011531445.1 2 - 3 293 135
XM_017005199.1 XP_016860688.1 3 - 4 520 135
XM_017005200.1 XP_016860689.1 3 - 4 1414 135
XM_017005201.1 XP_016860690.1 2 - 3 305 135
XM_017005202.1 XP_016860691.1 3 - 4 480 135
XM_017005203.1 XP_016860692.1 2 - 3 298 135
XM_017005204.1 XP_016860693.1 3 - 4 379 135
XM_017005205.1 XP_016860694.1 3 - 4 406 135
XM_017005206.1 XP_016860695.1 2 - 3 275 135
XM_017005207.1 XP_016860696.1 2 - 3 216 135
XM_017005208.1 XP_016860697.1 3 - 4 400 135
XM_017005209.1 XP_016860698.1 3 - 4 406 135
XM_017005210.1 XP_016860699.1 2 - 3 1227 135
XM_017005211.1 XP_016860700.1 3 - 4 453 135
XM_017005212.1 XP_016860701.1 2 - 3 1227 135
XM_017005214.1 XP_016860703.1 2 - 3 293 135
XM_017005215.1 XP_016860704.1 3 - 4 400 135
GenBank mRNA AB018356.1 - 2 - 3 322 135
AF105026.1 - 2 - 3 276 135
AF119415.1 - 2 - 3 241 135
AK001340.1 - 2 - 3 299 135
AK295252.1 - 2 - 3 216 135
AK315955.1 - 2 - 3 219 135
AY152815.2 - 2 - 3 197 135
AY359105.1 - 2 - 3 202 135
BC065936.1 - 2 - 3 258 135
BI820835.1 - 2 - 3 342 135
KU178447.1 - 1 - 2 129 135
KU178448.1 - 1 - 2 129 135

View Full Product Details