Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL5
Entrez Gene ID
8869
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
SATI, SIAT9, SIATGM3S, ST3GalV
Chromosome Location
Chr.2: 85839148 - 85889034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042437.1 NP_001035902.1 3 - 4 303 145
NM_003896.3 NP_003887.3 3 - 4 448 145
XM_005264630.4 XP_005264687.1 3 - 4 404 145
XM_017005199.1 XP_016860688.1 4 - 5 626 145
XM_017005200.1 XP_016860689.1 4 - 5 1520 145
XM_017005201.1 XP_016860690.1 3 - 4 411 145
XM_017005202.1 XP_016860691.1 4 - 5 586 145
GenBank mRNA AB018356.1 - 3 - 5 145
AF105026.1 - 3 - 4 382 145
AF119415.1 - 3 - 5 145
AK001340.1 - 3 - 4 405 145
AY152815.2 - 3 - 4 303 145
AY359105.1 - 3 - 4 308 145
BC065936.1 - 3 - 5 145
BI820835.1 - 3 - 5 145
KU178447.1 - 2 - 3 235 145
KU178448.1 - 2 - 3 235 145

View Full Product Details