Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
Chromosome Location
Chr.11: 95790461 - 95832693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243776.1 NP_001230705.1 8 - 9 1089 142
NM_014679.4 NP_055494.2 7 - 8 1045 142
XM_006718945.2 XP_006719008.1 6 - 7 928 142
XM_017018592.1 XP_016874081.1 8 - 9 2235 142
GenBank mRNA AK098647.1 - 7 - 8 1018 142
AK291245.1 - 7 - 8 1000 142
AK293277.1 - 8 - 9 1042 142
AY225092.1 - 7 - 8 807 142
AY239292.1 - 7 - 8 946 142
BC001233.2 - 7 - 8 1028 142
BC031048.1 - 7 - 8 956 142

View Full Product Details