Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM27
Entrez Gene ID
5987
Gene Name
tripartite motif containing 27
Gene Aliases
RFP, RNF76
Chromosome Location
Chr.6: 28903002 - 28923991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF230394.1 - 8 - 9 1291 104
AX776011.1 - 8 - 9 1415 104

View Full Product Details