The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022963.3 | NP_075252.2 | 7 - 8 | 1108 | 84 |
GenBank mRNA | AF202063.1 | - | 7 - 8 | 1091 | 84 |