The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000326.4 | NP_000317.1 | 7 - 8 | 1069 | 117 |
XM_011521870.2 | XP_011520172.1 | 7 - 8 | 2498 | 117 | |
XM_017022460.1 | XP_016877949.1 | 4 - 5 | 908 | 117 | |
GenBank mRNA | AK312457.1 | - | 7 - 8 | 957 | 117 |
BC004199.2 | - | 7 - 8 | 971 | 117 | |
BG205561.1 | - | 1 - 2 | 164 | 117 | |
J04213.1 | - | 6 - 7 | 872 | 117 |