Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLBP1
Entrez Gene ID
6017
Gene Name
retinaldehyde binding protein 1
Gene Aliases
CRALBP
Chromosome Location
Chr.15: 89209867 - 89223179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000326.4 NP_000317.1 8 - 9 1169 95
XM_011521870.2 XP_011520172.1 8 - 9 2598 95
XM_017022460.1 XP_016877949.1 5 - 6 1008 95
GenBank mRNA AK312457.1 - 8 - 9 1057 95
BC004199.2 - 8 - 9 1071 95
BG205561.1 - 2 - 3 264 95
BM704704.1 - 2 - 3 176 95
J04213.1 - 7 - 8 972 95

View Full Product Details