The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017775.3 | NP_060245.3 | 2 - 3 | 776 | 72 |
XM_017024801.1 | XP_016880290.1 | 2 - 3 | 689 | 72 | |
XM_017024802.1 | XP_016880291.1 | 2 - 3 | 689 | 72 | |
GenBank mRNA | AK056878.1 | - | 2 - 3 | 688 | 72 |
AK297783.1 | - | 2 - 3 | 332 | 72 | |
BC011698.2 | - | 2 - 3 | 315 | 72 | |
BC073796.1 | - | 2 - 3 | 319 | 72 | |
BC105128.1 | - | 2 - 3 | 776 | 72 | |
BC112107.1 | - | 2 - 3 | 776 | 72 |