The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271420.1 | NP_001258349.1 | 7 - 8 | 1278 | 110 |
NM_017775.3 | NP_060245.3 | 7 - 8 | 1141 | 110 | |
XM_017024801.1 | XP_016880290.1 | 7 - 8 | 1054 | 110 | |
XM_017024802.1 | XP_016880291.1 | 7 - 8 | 1054 | 110 | |
GenBank mRNA | AK000350.1 | - | 5 - 6 | 357 | 110 |
AK055780.1 | - | 7 - 8 | 834 | 110 | |
AK056878.1 | - | 7 - 8 | 1053 | 110 | |
AK297783.1 | - | 7 - 8 | 697 | 110 | |
BC011698.2 | - | 7 - 8 | 680 | 110 | |
BC073796.1 | - | 7 - 8 | 684 | 110 | |
BC105128.1 | - | 7 - 8 | 1141 | 110 | |
BC112107.1 | - | 7 - 8 | 1141 | 110 |