Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
Chromosome Location
Chr.16: 87406246 - 87492948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015144.2 NP_055959.1 6 - 7 659 91
XM_005255858.3 XP_005255915.2 6 - 7 850 91
XM_011522964.2 XP_011521266.1 6 - 7 1753 91
XM_017023082.1 XP_016878571.1 6 - 7 1609 91
GenBank mRNA AB011151.2 - 6 - 7 517 91
AB030243.1 - 6 - 7 667 91
AK289682.1 - 6 - 7 850 91
AL117532.1 - 3 - 4 282 91
BC101478.1 - 6 - 7 666 91

View Full Product Details