The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042749.2 | NP_001036214.1 | 30 - 31 | 3752 | 67 |
NM_001042750.1 | NP_001036215.1 | 30 - 31 | 3596 | 67 | |
NM_001042751.1 | NP_001036216.1 | 30 - 31 | 3596 | 67 | |
NM_001282418.1 | NP_001269347.1 | 30 - 31 | 3575 | 67 | |
NM_006603.4 | NP_006594.3 | 29 - 30 | 3531 | 67 | |
XM_005262357.2 | XP_005262414.1 | 29 - 30 | 3621 | 67 | |
XM_005262358.2 | XP_005262415.1 | 30 - 31 | 3558 | 67 | |
XM_005262359.3 | XP_005262416.1 | 29 - 30 | 3511 | 67 | |
XM_005262360.2 | XP_005262417.1 | 29 - 30 | 3440 | 67 | |
XM_005262361.2 | XP_005262418.1 | 29 - 30 | 3568 | 67 | |
XM_006724727.1 | XP_006724790.1 | 31 - 32 | 3700 | 67 | |
XM_006724728.3 | XP_006724791.1 | 30 - 31 | 3634 | 67 | |
XM_011531253.2 | XP_011529555.1 | 29 - 30 | 3493 | 67 | |
XM_017029232.1 | XP_016884721.1 | 30 - 31 | 3558 | 67 | |
XM_017029233.1 | XP_016884722.1 | 29 - 30 | 3440 | 67 | |
XM_017029234.1 | XP_016884723.1 | 29 - 30 | 3493 | 67 | |
XM_017029235.1 | XP_016884724.1 | 30 - 31 | 3639 | 67 | |
GenBank mRNA | AK298633.1 | - | 19 - 20 | 2250 | 67 |
AK313559.1 | - | 29 - 30 | 3423 | 67 | |
AL831939.2 | - | 29 - 30 | 3499 | 67 | |
BC001765.1 | - | 29 - 30 | 3480 | 67 | |
BX640970.1 | - | 30 - 31 | 3607 | 67 | |
BX641002.1 | - | 30 - 31 | 3500 | 67 | |
BX641003.1 | - | 30 - 31 | 3565 | 67 | |
CR627425.1 | - | 30 - 31 | 3574 | 67 | |
CR749433.1 | - | 30 - 31 | 3565 | 67 |