Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- STAG2
- Entrez Gene ID
-
10735
- Gene Name
- stromal antigen 2
- Gene Aliases
-
SA-2, SA2, SCC3B, bA517O1.1
- Chromosome Location
-
Chr.X: 123960560 - 124102656 on Build GRCh38
View Full Product Details