The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165966.1 | NP_001159438.1 | 16 - 17 | 2345 | 90 |
NM_031220.3 | NP_112497.2 | 17 - 18 | 2453 | 90 | |
XM_011524015.2 | XP_011522317.1 | 17 - 18 | 2393 | 90 | |
GenBank mRNA | AB209130.1 | - | 11 - 12 | 4816 | 90 |
AF334586.1 | - | 17 - 18 | 2450 | 90 | |
AL389994.1 | - | 16 - 17 | 2176 | 90 | |
BC020484.1 | - | 8 - 9 | 1051 | 90 | |
BC128583.1 | - | 17 - 18 | 2390 | 90 | |
BC128584.1 | - | 17 - 18 | 2390 | 90 |