The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130480.2 | NP_001123952.1 | 11 - 12 | 1209 | 109 |
NM_001130481.2 | NP_001123953.1 | 11 - 12 | 1051 | 109 | |
NM_001130482.2 | NP_001123954.1 | 11 - 12 | 1155 | 109 | |
NM_001130483.2 | NP_001123955.1 | 11 - 12 | 1209 | 109 | |
NM_001288836.1 | NP_001275765.1 | 12 - 13 | 1346 | 109 | |
NM_001288837.1 | NP_001275766.1 | 14 - 15 | 1769 | 109 | |
NM_001288838.1 | NP_001275767.1 | 13 - 14 | 1651 | 109 | |
NM_001288839.1 | NP_001275768.1 | 10 - 11 | 1186 | 109 | |
NM_006296.6 | NP_006287.2 | 11 - 12 | 1256 | 109 | |
XM_005264540.4 | XP_005264597.1 | 11 - 12 | 1269 | 109 | |
XM_006712090.3 | XP_006712153.1 | 11 - 12 | 1064 | 109 | |
XM_006712091.3 | XP_006712154.1 | 11 - 12 | 1304 | 109 | |
XM_006712092.3 | XP_006712155.1 | 12 - 13 | 1441 | 109 | |
XM_006712093.3 | XP_006712156.1 | 9 - 10 | 1146 | 109 | |
XM_011533092.2 | XP_011531394.1 | 11 - 12 | 1053 | 109 | |
XM_017004858.1 | XP_016860347.1 | 11 - 12 | 1209 | 109 | |
XM_017004859.1 | XP_016860348.1 | 12 - 13 | 1931 | 109 | |
XM_017004860.1 | XP_016860349.1 | 11 - 12 | 1861 | 109 | |
XM_017004861.1 | XP_016860350.1 | 9 - 10 | 960 | 109 | |
XM_017004862.1 | XP_016860351.1 | 10 - 11 | 1773 | 109 | |
XM_017004863.1 | XP_016860352.1 | 11 - 12 | 1353 | 109 | |
XM_017004864.1 | XP_016860353.1 | 12 - 13 | 1441 | 109 | |
XM_017004865.1 | XP_016860354.1 | 12 - 13 | 1255 | 109 | |
XM_017004866.1 | XP_016860355.1 | 12 - 13 | 1304 | 109 | |
GenBank mRNA | AB000450.1 | - | 11 - 12 | 1154 | 109 |
AJ512204.2 | - | 11 - 12 | 1154 | 109 | |
AK058199.1 | - | 11 - 12 | 1111 | 109 | |
AK223540.1 | - | 11 - 12 | 1111 | 109 | |
AK292727.1 | - | 13 - 14 | 1637 | 109 | |
AK296611.1 | - | 12 - 13 | 1331 | 109 | |
AK311205.1 | - | 1 - 2 | 91 | 109 | |
AY228367.1 | - | 11 - 12 | 1051 | 109 | |
AY228368.1 | - | 11 - 12 | 1186 | 109 | |
AY228369.1 | - | 11 - 12 | 1084 | 109 | |
AY228370.1 | - | 12 - 13 | 1276 | 109 | |
AY228371.1 | - | 10 - 11 | 1116 | 109 | |
AY228372.1 | - | 11 - 12 | 1139 | 109 | |
BC021663.1 | - | 11 - 12 | 1082 | 109 | |
BC027854.1 | - | 11 - 12 | 1207 | 109 |