Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VRK2
Entrez Gene ID
7444
Gene Name
vaccinia related kinase 2
Gene Aliases
-
Chromosome Location
Chr.2: 57907651 - 58163991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130480.2 NP_001123952.1 5 - 6 530 103
NM_001130481.2 NP_001123953.1 5 - 6 372 103
NM_001130482.2 NP_001123954.1 5 - 6 476 103
NM_001130483.2 NP_001123955.1 5 - 6 530 103
NM_001288836.1 NP_001275765.1 6 - 7 667 103
NM_001288837.1 NP_001275766.1 8 - 9 1090 103
NM_001288838.1 NP_001275767.1 7 - 8 972 103
NM_001288839.1 NP_001275768.1 4 - 5 507 103
NM_006296.6 NP_006287.2 5 - 6 577 103
XM_005264540.4 XP_005264597.1 5 - 6 590 103
XM_006712090.3 XP_006712153.1 5 - 6 385 103
XM_006712091.3 XP_006712154.1 5 - 6 625 103
XM_006712092.3 XP_006712155.1 6 - 7 762 103
XM_011533092.2 XP_011531394.1 5 - 6 374 103
XM_017004858.1 XP_016860347.1 5 - 6 530 103
XM_017004859.1 XP_016860348.1 6 - 7 1252 103
XM_017004860.1 XP_016860349.1 5 - 6 1182 103
XM_017004863.1 XP_016860352.1 5 - 6 674 103
XM_017004864.1 XP_016860353.1 6 - 7 762 103
XM_017004865.1 XP_016860354.1 6 - 7 576 103
XM_017004866.1 XP_016860355.1 6 - 7 625 103
GenBank mRNA AB000450.1 - 5 - 6 475 103
AJ512204.2 - 5 - 6 475 103
AK058199.1 - 5 - 6 432 103
AK223540.1 - 5 - 6 432 103
AK292727.1 - 7 - 8 958 103
AK295193.1 - 5 - 6 422 103
AK296611.1 - 6 - 7 652 103
AY228367.1 - 5 - 6 372 103
AY228368.1 - 5 - 6 507 103
AY228369.1 - 5 - 6 405 103
AY228370.1 - 6 - 7 597 103
AY228371.1 - 4 - 5 437 103
AY228372.1 - 5 - 6 460 103
BC021663.1 - 5 - 6 403 103
BC027854.1 - 5 - 6 528 103

View Full Product Details