Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFAF7
Entrez Gene ID
55471
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
C2orf56, MidA, PRO1853
Chromosome Location
Chr.2: 37231631 - 37259052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144736.4 NP_653337.1 2 - 3 289 65
XM_005264410.1 XP_005264467.1 2 - 3 269 65
XM_005264411.1 XP_005264468.1 2 - 3 267 65
XM_011532956.1 XP_011531258.1 2 - 3 257 65
GenBank mRNA AK294240.1 - 2 - 3 223 65
AK299015.1 - 2 - 3 263 65
AK303555.1 - 2 - 3 269 65
BC004548.2 - 2 - 3 254 65
BI334629.1 - 2 - 3 240 65
BM172474.1 - 2 - 3 294 65

View Full Product Details