The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286834.1 | NP_001273763.1 | 5 - 6 | 676 | 145 |
NM_001286837.1 | NP_001273766.1 | 5 - 6 | 914 | 145 | |
NM_002936.4 | NP_002927.2 | 5 - 6 | 676 | 145 | |
GenBank mRNA | AF039652.1 | - | 5 - 6 | 646 | 145 |
AF048994.1 | - | 5 - 6 | 592 | 145 | |
AF048995.1 | - | 5 - 6 | 626 | 145 | |
AK075490.1 | - | 5 - 6 | 625 | 145 | |
AK096913.1 | - | 5 - 6 | 832 | 145 | |
BC002973.1 | - | 5 - 6 | 635 | 145 | |
BT019670.1 | - | 5 - 6 | 565 | 145 | |
CR541820.1 | - | 5 - 6 | 565 | 145 |