Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKD2
Entrez Gene ID
85409
Gene Name
naked cuticle homolog 2
Gene Aliases
Naked2
Chromosome Location
Chr.5: 1008845 - 1038812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271082.1 NP_001258011.1 9 - 10 880 72
XM_005248383.3 XP_005248440.1 9 - 10 1144 72
GenBank mRNA BC012176.1 - 9 - 10 880 72

View Full Product Details