The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005273613.3 | XP_005273670.1 | 1 - 2 | 167 | 59 |
XM_005273615.3 | XP_005273672.1 | 1 - 2 | 167 | 59 | |
XM_006716390.3 | XP_006716453.1 | 1 - 2 | 167 | 59 | |
XM_017013750.1 | XP_016869239.1 | 1 - 2 | 167 | 59 |