Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNN1
Entrez Gene ID
3780
Gene Name
potassium calcium-activated channel subfamily N member 1
Gene Aliases
KCa2.1, SK1, SKCA1, hSK1
Chromosome Location
Chr.19: 17951302 - 17999121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002248.4 NP_002239.2 1 - 2 107 96
GenBank mRNA U69883.2 - 1 - 2 116 96

View Full Product Details