Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FANCM
Entrez Gene ID
57697
Gene Name
Fanconi anemia complementation group M
Gene Aliases
FAAP250, KIAA1596
Chromosome Location
Chr.14: 45135939 - 45200890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK001672.1 - 7 - 8 2137 95

View Full Product Details