The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007192.3 | NP_009123.1 | 22 - 23 | 3004 | 72 |
XM_011536381.2 | XP_011534683.1 | 23 - 24 | 3006 | 72 | |
GenBank mRNA | AF164924.1 | - | 2 - 3 | 265 | 72 |
AK001719.1 | - | 22 - 23 | 2740 | 72 | |
AK091676.1 | - | 2 - 3 | 651 | 72 |