The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007192.3 | NP_009123.1 | 8 - 9 | 1389 | 70 |
XM_011536381.2 | XP_011534683.1 | 9 - 10 | 1391 | 70 | |
GenBank mRNA | AK001719.1 | - | 8 - 9 | 1125 | 70 |
AK024072.1 | - | 8 - 9 | 1391 | 70 | |
BC000565.2 | - | 8 - 9 | 1126 | 70 | |
BC014046.2 | - | 8 - 9 | 1126 | 70 | |
BC064561.1 | - | 8 - 9 | 1114 | 70 | |
BC073849.1 | - | 8 - 9 | 1123 | 70 | |
BC111402.1 | - | 8 - 9 | 1138 | 70 |