Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRY
Entrez Gene ID
10129
Gene Name
FRY microtubule binding protein
Gene Aliases
13CDNA73, 214K23.2, C13orf14, CG003, bA207N4.2, bA37E23.1
Chromosome Location
Chr.13: 32029742 - 32299122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023037.2 NP_075463.2 7 - 8 1215 85
XM_006719749.3 XP_006719812.1 7 - 8 2296 85
XM_017020303.1 XP_016875792.1 7 - 8 2773 85
XM_017020304.1 XP_016875793.1 7 - 8 1215 85
XM_017020305.1 XP_016875794.1 7 - 8 2792 85
GenBank mRNA AL049784.1 - 7 - 8 1215 85
U50534.1 - 7 - 8 1214 85

View Full Product Details