The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004685.3 | NP_004676.3 | 2 - 3 | 461 | 122 |
XM_011535306.1 | XP_011533608.1 | 2 - 3 | 407 | 122 | |
XM_011535307.1 | XP_011533609.1 | 2 - 3 | 407 | 122 | |
XM_017020845.1 | XP_016876334.1 | 2 - 3 | 407 | 122 | |
GenBank mRNA | AF406619.1 | - | 2 - 3 | 142 | 122 |
AK314587.1 | - | 2 - 3 | 326 | 122 | |
AL832017.2 | - | 2 - 3 | 384 | 122 | |
BC040012.1 | - | 2 - 3 | 333 | 122 |