Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
-
Chromosome Location
Chr.13: 25246201 - 25287566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004685.3 NP_004676.3 9 - 10 1409 114
XM_011535306.1 XP_011533608.1 10 - 11 1469 114
XM_011535307.1 XP_011533609.1 10 - 11 1469 114
XM_017020845.1 XP_016876334.1 9 - 10 1355 114
XM_017020846.1 XP_016876335.1 6 - 7 674 114
GenBank mRNA AF072928.1 - 5 - 6 623 114
AF406619.1 - 9 - 10 1090 114
AL832017.2 - 9 - 10 1332 114
BC040012.1 - 9 - 10 1281 114
BU177677.1 - 4 - 5 474 114

View Full Product Details