Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MASP1
Entrez Gene ID
5648
Gene Name
mannan binding lectin serine peptidase 1
Gene Aliases
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF
Chromosome Location
Chr.3: 187216085 - 187292220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031849.2 NP_001027019.1 7 - 8 1398 90
NM_001879.5 NP_001870.3 7 - 8 1398 90
NM_139125.3 NP_624302.1 7 - 8 1398 90
NR_033519.1 6 - 7 1166 90
XM_006713701.2 XP_006713764.1 8 - 9 1471 90
XM_011512989.2 XP_011511291.1 7 - 8 1392 90
XM_011512990.2 XP_011511292.1 7 - 8 1392 90
XM_011512991.2 XP_011511293.1 7 - 8 1387 90
XM_017006869.1 XP_016862358.1 8 - 9 1476 90
XM_017006870.1 XP_016862359.1 6 - 7 1364 90
XM_017006871.1 XP_016862360.1 7 - 8 1392 90
XM_017006872.1 XP_016862361.1 7 - 8 1251 90
GenBank mRNA AF284421.1 - 7 - 8 1098 90
AK129893.1 - 7 - 8 1097 90
AK291157.1 - 7 - 8 1234 90
AK291686.1 - 7 - 8 1076 90
AK300663.1 - 8 - 9 1219 90
AK304334.1 - 6 - 7 876 90
BC039724.1 - 7 - 8 1333 90
BC106945.1 - 7 - 8 1058 90
BC106946.1 - 7 - 8 1058 90
BX647237.1 - 6 - 7 881 90
CR749615.1 - 7 - 8 1300 90

View Full Product Details