The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015078.3 | NP_055893.3 | 17 - 18 | 2337 | 141 |
XM_011512585.2 | XP_011510887.1 | 9 - 10 | 1774 | 141 | |
XM_011512586.2 | XP_011510888.1 | 10 - 11 | 1188 | 141 | |
XM_017005943.1 | XP_016861432.1 | 17 - 18 | 3058 | 141 | |
GenBank mRNA | AB020668.1 | - | 14 - 15 | 1649 | 141 |
AY172737.1 | - | 16 - 17 | 1889 | 141 | |
BC064632.1 | - | 17 - 18 | 2153 | 141 | |
BX649003.1 | - | - | 2387 | 141 |