The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015078.3 | NP_055893.3 | 25 - 26 | 3178 | 114 |
XM_011512585.2 | XP_011510887.1 | 17 - 18 | 2615 | 114 | |
XM_011512586.2 | XP_011510888.1 | 18 - 19 | 2029 | 114 | |
GenBank mRNA | AB020668.1 | - | 22 - 23 | 2490 | 114 |
AY172737.1 | - | 24 - 25 | 2730 | 114 | |
BC029074.2 | - | 4 - 5 | 479 | 114 | |
BX649003.1 | - | - | 7357 | 114 |