The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276342.1 | NP_001263271.1 | 14 - 15 | 1493 | 91 |
NM_001318707.1 | NP_001305636.1 | 13 - 14 | 1408 | 91 | |
NM_001318708.1 | NP_001305637.1 | 15 - 16 | 1552 | 91 | |
NM_001318709.1 | NP_001305638.1 | 14 - 15 | 1493 | 91 | |
NM_013326.4 | NP_037458.3 | 16 - 17 | 1637 | 91 | |
XM_011525939.1 | XP_011524241.1 | 15 - 16 | 1509 | 91 | |
XM_017025717.1 | XP_016881206.1 | 14 - 15 | 1446 | 91 | |
GenBank mRNA | AF143536.1 | - | 15 - 16 | 1420 | 91 |
AK027106.1 | - | 8 - 9 | 712 | 91 | |
AK057192.1 | - | 16 - 17 | 1598 | 91 | |
AK294169.1 | - | 13 - 14 | 1355 | 91 | |
AK295246.1 | - | 15 - 16 | 1509 | 91 | |
AK296230.1 | - | 14 - 15 | 1444 | 91 | |
BC002950.1 | - | 16 - 17 | 1562 | 91 | |
BC008305.1 | - | 16 - 17 | 1569 | 91 |