The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018227.5 | NP_060697.4 | 15 - 16 | 1372 | 77 |
XM_017008359.1 | XP_016863848.1 | 15 - 16 | 1349 | 77 | |
GenBank mRNA | AB014773.1 | - | 15 - 16 | 1320 | 77 |
AK094164.1 | - | 7 - 8 | 578 | 77 | |
AK314371.1 | - | 15 - 16 | 1349 | 77 | |
AL832015.2 | - | 15 - 16 | 1341 | 77 | |
AL832458.2 | - | 15 - 16 | 1341 | 77 | |
AY359880.1 | - | 2 - 3 | 693 | 77 | |
BC126484.1 | - | 15 - 16 | 1367 | 77 | |
BC126486.1 | - | 15 - 16 | 1367 | 77 | |
EF623993.1 | - | 15 - 16 | 1320 | 77 |