The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018227.5 | NP_060697.4 | 19 - 20 | 1790 | 154 |
XM_017008359.1 | XP_016863848.1 | 19 - 20 | 1767 | 154 | |
GenBank mRNA | AB014773.1 | - | 19 - 20 | 1738 | 154 |
AK001670.1 | - | 1 - 2 | 139 | 154 | |
AK094164.1 | - | 11 - 12 | 996 | 154 | |
AK314371.1 | - | 19 - 20 | 1767 | 154 | |
AL832015.2 | - | 19 - 20 | 1759 | 154 | |
AL832458.2 | - | 19 - 20 | 1759 | 154 | |
AY359880.1 | - | 6 - 7 | 1111 | 154 | |
BC126484.1 | - | 19 - 20 | 1785 | 154 | |
BC126486.1 | - | 19 - 20 | 1785 | 154 | |
EF623993.1 | - | 19 - 20 | 1738 | 154 |