The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018227.5 | NP_060697.4 | 3 - 4 | 284 | 62 |
XM_017008359.1 | XP_016863848.1 | 3 - 4 | 261 | 62 | |
GenBank mRNA | AB014773.1 | - | 3 - 4 | 232 | 62 |
AK094969.1 | - | 3 - 4 | 261 | 62 | |
AK314371.1 | - | 3 - 4 | 261 | 62 | |
AL596839.1 | - | 3 - 4 | 312 | 62 | |
AL832015.2 | - | 3 - 4 | 253 | 62 | |
AL832458.2 | - | 3 - 4 | 253 | 62 | |
BC031637.1 | - | 3 - 4 | 266 | 62 | |
BC126484.1 | - | 3 - 4 | 279 | 62 | |
BC126486.1 | - | 3 - 4 | 279 | 62 | |
EF623993.1 | - | 3 - 4 | 232 | 62 |