The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165136.1 | NP_001158608.1 | 17 - 18 | 2496 | 100 |
NM_017912.3 | NP_060382.3 | 18 - 19 | 2604 | 100 | |
XM_005263083.3 | XP_005263140.1 | 18 - 19 | 2801 | 100 | |
XM_011532053.2 | XP_011530355.1 | 17 - 18 | 2695 | 100 | |
XM_017008333.1 | XP_016863822.1 | 17 - 18 | 2162 | 100 | |
GenBank mRNA | AF336798.1 | - | 18 - 19 | 2604 | 100 |
AK000644.1 | - | 5 - 6 | 692 | 100 | |
AK225506.1 | - | 5 - 6 | 692 | 100 | |
AK295832.1 | - | 18 - 19 | 2514 | 100 | |
AL833664.1 | - | 7 - 8 | 932 | 100 | |
AY653201.1 | - | 17 - 18 | 2496 | 100 | |
AY653202.1 | - | 17 - 18 | 2408 | 100 | |
AY653203.1 | - | 15 - 16 | 2189 | 100 | |
BC035775.1 | - | 4 - 5 | 544 | 100 | |
BC042047.1 | - | 18 - 19 | 2480 | 100 | |
BX647121.1 | - | 18 - 19 | 2280 | 100 |