Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
Chromosome Location
Chr.14: 57202476 - 57268899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006544.3 NP_006535.1 12 - 13 1552 159
XM_005267272.3 XP_005267329.1 12 - 13 1637 159
GenBank mRNA AK295085.1 - 12 - 13 1459 159
AK303531.1 - 11 - 12 1295 159
AK312523.1 - 12 - 13 1557 159
AL833745.1 - 2 - 3 1019 159
BC041126.1 - 12 - 13 1518 159
U85946.1 - 12 - 13 1496 159

View Full Product Details