Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
Chromosome Location
Chr.14: 57202476 - 57268899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006544.3 NP_006535.1 16 - 17 1980 151
XM_005267272.3 XP_005267329.1 16 - 17 2065 151
GenBank mRNA AK295085.1 - 16 - 17 1887 151
AK303531.1 - 15 - 16 1723 151
AK312523.1 - 16 - 17 1985 151
AL833745.1 - 6 - 7 1447 151
BC041126.1 - 16 - 17 1946 151
U85946.1 - 16 - 17 1924 151

View Full Product Details